MilliporeSigma
  • A facile stable-isotope dilution method for determination of sphingosine phosphate lyase activity.
A facile stable-isotope dilution method for determination of sphingosine phosphate lyase activity.
Chemistry and physics of lipids (2015-09-27)
Jung H Suh, Abeer Eltanawy, Apoorva Rangan, Julie D Saba
ABSTRACT
A new technique for quantifying sphingosine phosphate lyase activity in biological samples is described. In this procedure, 2-hydrazinoquinoline is used to convert (2E)-hexadecenal into the corresponding hydrazone derivative to improve ionization efficiency and selectivity of detection. Combined utilization of liquid chromatographic separation and multiple reaction monitoring-mass spectrometry allows for simultaneous quantification of the substrate S1P and product (2E)-hexadecenal. Incorporation of (2E)- d5-hexadecenal as an internal standard improves detection accuracy and precision. A simple one-step derivatization procedure eliminates the need for further extractions. Limits of quantification for (2E)-hexadecenal and sphingosine-1-phosphate are 100 and 50fmol, respectively. The assay displays a wide dynamic detection range useful for detection of low basal sphingosine phosphate lyase activity in wild type cells, SPL-overexpressing cell lines, and wild type mouse tissues. Compared to current methods, the capacity for simultaneous detection of sphingosine-1-phosphate and (2E)-hexadecenal greatly improves the accuracy of results and shows excellent sensitivity and specificity for sphingosine phosphate lyase activity detection.