MilliporeSigma
  • Orthogonal Genetic Regulation in Human Cells Using Chemically Induced CRISPR/Cas9 Activators.
Orthogonal Genetic Regulation in Human Cells Using Chemically Induced CRISPR/Cas9 Activators.
ACS synthetic biology (2017-01-06)
Zehua Bao, Surbhi Jain, Valerie Jaroenpuntaruk, Huimin Zhao
ABSTRACT
The concerted action of multiple genes in a time-dependent manner controls complex cellular phenotypes, yet the temporal regulation of gene expressions is restricted on a single-gene level, which limits our ability to control higher-order gene networks and understand the consequences of multiplex genetic perturbations. Here we developed a system for temporal regulation of multiple genes. This system combines the simplicity of CRISPR/Cas9 activators for orthogonal targeting of multiple genes and the orthogonality of chemically induced dimerizing (CID) proteins for temporal control of CRISPR/Cas9 activator function. In human cells, these transcription activators exerted simultaneous activation of multiple genes and orthogonal regulation of different genes in a ligand-dependent manner with minimal background. We envision that our system will enable the perturbation of higher-order gene networks with high temporal resolution and accelerate our understanding of gene-gene interactions in a complex biological setting.
Sigma-Aldrich
GA3-AM, ≥95% (HPLC)