MilliporeSigma
  • Epstein-Barr virus-encoded latent membrane protein 1 induces epithelial to mesenchymal transition by inducing V-set Ig domain containing 4 (VSIG4) expression via NF-kB in renal tubular epithelial HK-2 cells.
Epstein-Barr virus-encoded latent membrane protein 1 induces epithelial to mesenchymal transition by inducing V-set Ig domain containing 4 (VSIG4) expression via NF-kB in renal tubular epithelial HK-2 cells.
Biochemical and biophysical research communications (2017-09-02)
Seung-Mi Kim, Se Won Oh, Sang Hyun Park, Dae Young Hur, Seung-Woo Hong, Sang Youb Han
ABSTRACT
The epithelial to mesenchymal transition (EMT), a hallmark of chronic kidney disease, is a key event in the conversion from tubular epithelial cells to myofibroblasts in renal fibrosis. Epstein-Barr virus (EBV) is a γ-herpes oncovirus associated with chronic kidney disease. However, the relationship between EBV and the EMT process in renal tubular epithelial cells is not well understood. Among EBV-latent genes, EBV-encoded latent membrane protein 1 (LMP1) induces EMT by regulating a variety of molecules in EBV-induced oncogenic transformation. In this study, we investigated EBV-encoded LMP1 and EMT process markers in human proximal tubule epithelial cell line HK-2. LMP1 overexpression induces cell morphological changes via the epithelial to mesenchymal process in HK-2 cells, and these changes accelerate cell proliferation, cell motility, and invasion. Furthermore, VSIG4 upregulation by EBV-LMP1 induced LMP1-mediated EMT, cell motility, and invasion. VSIG4 upregulation by LMP1 was regulated at the transcriptional level via the NF-kB signaling axis. These results suggest that EBV-encoded LMP1 regulates EMT through the NF-kB-VSIG4 axis in HK-2 cells, and VSIG4 is a potential target in EBV-induced chronic kidney diseases.
Sigma-Aldrich
MISSION® esiRNA, targeting human VSIG4