MilliporeSigma
  • The nuclear receptor ERβ engages AGO2 in regulation of gene transcription, RNA splicing and RISC loading.
The nuclear receptor ERβ engages AGO2 in regulation of gene transcription, RNA splicing and RISC loading.
Genome biology (2017-10-12)
Roberta Tarallo, Giorgio Giurato, Giuseppina Bruno, Maria Ravo, Francesca Rizzo, Annamaria Salvati, Luca Ricciardi, Giovanna Marchese, Angela Cordella, Teresa Rocco, Valerio Gigantino, Biancamaria Pierri, Giovanni Cimmino, Luciano Milanesi, Concetta Ambrosino, Tuula A Nyman, Giovanni Nassa, Alessandro Weisz
ABSTRACT
The RNA-binding protein Argonaute 2 (AGO2) is a key effector of RNA-silencing pathways It exerts a pivotal role in microRNA maturation and activity and can modulate chromatin remodeling, transcriptional gene regulation and RNA splicing. Estrogen receptor beta (ERβ) is endowed with oncosuppressive activities, antagonizing hormone-induced carcinogenesis and inhibiting growth and oncogenic functions in luminal-like breast cancers (BCs), where its expression correlates with a better prognosis of the disease. Applying interaction proteomics coupled to mass spectrometry to characterize nuclear factors cooperating with ERβ in gene regulation, we identify AGO2 as a novel partner of ERβ in human BC cells. ERβ-AGO2 association was confirmed in vitro and in vivo in both the nucleus and cytoplasm and is shown to be RNA-mediated. ChIP-Seq demonstrates AGO2 association with a large number of ERβ binding sites, and total and nascent RNA-Seq in ERβ + vs ERβ - cells, and before and after AGO2 knock-down in ERβ + cells, reveals a widespread involvement of this factor in ERβ-mediated regulation of gene transcription rate and RNA splicing. Moreover, isolation and sequencing by RIP-Seq of ERβ-associated long and small RNAs in the cytoplasm suggests involvement of the nuclear receptor in RISC loading, indicating that it may also be able to directly control mRNA translation efficiency and stability. These results demonstrate that AGO2 can act as a pleiotropic functional partner of ERβ, indicating that both factors are endowed with multiple roles in the control of key cellular functions.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Millipore
EZview Red Anti-c-Myc Affinity Gel
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse
Sigma-Aldrich
Anti-Ago 2 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Anti-Ago1 Antibody, clone 4G7-E12, clone 4G7-E12, from mouse