MilliporeSigma
  • Small and Intermediate Calcium-Activated Potassium Channel Openers Improve Rat Endothelial and Erectile Function.
Small and Intermediate Calcium-Activated Potassium Channel Openers Improve Rat Endothelial and Erectile Function.
Frontiers in pharmacology (2017-10-11)
Simon G Comerma-Steffensen, Ingrid Carvacho, Elise R Hedegaard, Ulf Simonsen
ABSTRACT
Modulation of endothelial calcium-activated potassium (K
Sigma-Aldrich
TRAM-34, ≥98% (HPLC), solid
Sigma-Aldrich
Linopirdine, ≥98% (HPLC)
Sigma-Aldrich
Acetylcholine bromide, ≥99%