MilliporeSigma
  • Soluble epoxide hydrolase inhibitor AUDA decreases bleomycin-induced pulmonary toxicity in mice by inhibiting the p38/Smad3 pathways.
Soluble epoxide hydrolase inhibitor AUDA decreases bleomycin-induced pulmonary toxicity in mice by inhibiting the p38/Smad3 pathways.
Toxicology (2017-07-12)
Xin-Wei Dong, Yong-Liang Jia, Ling-Tian Ge, Bo Jiang, Jun-Xia Jiang, Jian Shen, Ya-Chao Jin, Yan Guan, Yun Sun, Qiang-Min Xie
ABSTRACT
Bleomycin (BLM) has potent tumor cell-killing properties that have given it an important place in cancer chemotherapy, but pulmonary toxicity is its major adverse effect. Soluble epoxide hydrolase (sEH) inhibitors have been reported to have protective effects in fibrosis models, but the effects of AUDA, an sEH inhibitor of BLM-induced pulmonary toxicity and fibrosis, remain to be researched. In this study, we assessed the effects of AUDA on the BLM-induced pulmonary fibrosis in a mouse model, and transforming growth factor (TGF)-β
Sigma-Aldrich
SIS3, ≥98% (HPLC), powder
Sigma-Aldrich
AUDA, ≥98% (HPLC)