MilliporeSigma
  • Quantification of vicine and convicine in faba bean seeds using hydrophilic interaction liquid chromatography.
Quantification of vicine and convicine in faba bean seeds using hydrophilic interaction liquid chromatography.
Food chemistry (2017-09-28)
Randy W Purves, Hamid Khazaei, Albert Vandenberg
ABSTRACT
Faba bean (Vicia faba L.) provides environmental and health benefits; however, the presence of the pyrimidine glycosides vicine and convicine (v-c) in its seeds limits consumption. Low v-c genotypes have been introduced, but the convicine levels in these genotypes have not been quantified. To improve detection, the polar nature of v-c was exploited by implementing hydrophilic interaction liquid chromatography (HILIC). A sample preparation method using a two-step extraction was developed for use with UV and/or tandem mass spectrometry (SRM) detection. The HILIC-UV method was suitable for over three orders of magnitude, covering the range of v-c concentrations in faba bean seeds across all genotypes tested. The linear range of HILIC-SRM was slightly less (∼3 orders of magnitude), but improved sensitivity and selectivity make it more suitable for quantifying low v-c samples. The analysis of 13 genotypes suggests that v-c concentrations in faba bean seeds may be independent quantitative traits.