MilliporeSigma
  • Low-carbohydrate diet induces metabolic depression: a possible mechanism to conserve glycogen.
Low-carbohydrate diet induces metabolic depression: a possible mechanism to conserve glycogen.
American journal of physiology. Regulatory, integrative and comparative physiology (2017-07-14)
Hugh S Winwood-Smith, Craig E Franklin, Craig R White
ABSTRACT
Long-term studies have found that low-carbohydrate diets are more effective for weight loss than calorie-restricted diets in the short term but equally or only marginally more effective in the long term. Low-carbohydrate diets have been linked to reduced glycogen stores and increased feelings of fatigue. We propose that reduced physical activity in response to lowered glycogen explains the diminishing weight loss advantage of low-carbohydrate compared with low-calorie diets over longer time periods. We explored this possibility by feeding adult
Product Description
Supelco
Bradford Reagent, for 0.1-1.4 mg/ml protein
Sigma-Aldrich
Amyloglucosidase from Aspergillus niger, ammonium sulfate suspension, ≥40 units/mg protein