MilliporeSigma
Generation of Persister Cells of
Frontiers in microbiology (2017-10-20)
Lucia Grassi, Mariagrazia Di Luca, Giuseppantonio Maisetta, Andrea C Rinaldi, Semih Esin, Andrej Trampuz, Giovanna Batoni
ABSTRACT
Persister cells (PCs) are a subset of dormant, phenotypic variants of regular bacteria, highly tolerant to antibiotics. Generation of PCs
Sigma-Aldrich
Daptomycin, cyclic lipopeptide antibiotic