MilliporeSigma
PEGylated (NH
Journal of photochemistry and photobiology. B, Biology (2017-07-28)
Daniel K Macharia, Qiyun Tian, Liang Chen, Yingqi Sun, Nuo Yu, Chuanglong He, Han Wang, Zhigang Chen
ABSTRACT
The simultaneous imaging and photothermal therapy of tumors have attracted much attention, and a prerequisite is to obtain multifunctional nanomaterials. Ideally, one kind of nanoparticles with single component can be used as both imaging agent and photothermal agent. Herein, we have developed the PEGylated (NH