MilliporeSigma
  • Wogonoside exerts growth-suppressive effects against T acute lymphoblastic leukemia through the STAT3 pathway.
Wogonoside exerts growth-suppressive effects against T acute lymphoblastic leukemia through the STAT3 pathway.
Human & experimental toxicology (2016-12-13)
R Xiao, M Gan, T Jiang
ABSTRACT
Wogonoside is the main flavonoid of the traditional Chinese medicinal herb Scutellaria baicalensis Georgi and has been found to induce growth suppression in myelogenous leukemia cells. However, its activity in T acute lymphoblastic leukemia (T-ALL) is still unclear. In this study, T-ALL cell lines MOLT-3 and Jurkat were exposed to different concentrations of wogonoside for 48 h, and cell viability, cell cycle distribution, and apoptosis were measured. The involvement of signal transducers and activators of transcription 3 (STAT3) signaling in the activity of wogonoside was checked. The in vivo effect of wogonoside on T-ALL growth was investigated in a xenograft mouse model. Wogonoside significantly inhibited the viability of MOLT-3 and Jurkat cells, with the IC
Sigma-Aldrich
Wogonoside, ≥95% (LC/MS-ELSD)