MilliporeSigma
  • Authenticity determination of honeys with non-extractable proteins by means of elemental analyzer (EA) and liquid chromatography (LC) coupled to isotope ratio mass spectroscopy (IRMS).
Authenticity determination of honeys with non-extractable proteins by means of elemental analyzer (EA) and liquid chromatography (LC) coupled to isotope ratio mass spectroscopy (IRMS).
Food chemistry (2017-09-28)
Hao Dong, Kaijun Xiao, Yanping Xian, Yuluan Wu
ABSTRACT
The present work aims to systematically demonstrate the authenticity of honeys with non-extractable proteins for the first time, by means of EA-IRMS and LC-IRMS. Fifty-three pure honeys of various botanical and geographical origins were studied and a criterion on the basis of the stable carbon isotope ratio characterization of total honey and the main sugars was established for pure honeys. Parameters such as δ
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O
Sigma-Aldrich
Isomaltose, ~98% (TLC)
Sigma-Aldrich
Phosphoric acid, puriss., ≥99%