MilliporeSigma
Strontium release from Sr
Journal of controlled release : official journal of the Controlled Release Society (2017-08-02)
Marcus Rohnke, Stefanie Pfitzenreuter, Boris Mogwitz, Anja Henß, Jürgen Thomas, Dina Bieberstein, Thomas Gemming, Svenja K Otto, Seemun Ray, Matthias Schumacher, Michael Gelinsky, Volker Alt
ABSTRACT
Drug functionalization of biomaterials is a modern and popular approach in biomaterials research. Amongst others this concept is used for the functionalization of bone implants to locally stimulate the bone healing process. For example strontium ions (Sr
Sigma-Aldrich
Minimum Essential Medium Eagle, Joklik Modification, With sodium bicarbonate, without L-glutamine, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Penicillin-Streptomycin, Solution stabilized, with 10,000 units penicillin and 10 mg streptomycin/mL, 0.1 μm filtered, BioReagent, suitable for cell culture