MilliporeSigma
A Mini Zinc-Finger Protein (MIF) from
Frontiers in plant science (2017-10-12)
Meixiang Han, Xuefeng Jin, Wei Yao, Lingjie Kong, Gan Huang, Yujin Tao, Lingfei Li, Xiaojing Wang, Yaqin Wang
ABSTRACT
Petal appearance is an important horticultural trail that is generally used to evaluate the ornamental value of plants. However, knowledge of the molecular regulation of petal growth is mostly derived from analyses of
Product Description
Supelco
α-tert-Butyl-β-(4-chlorobenzyl)-1H-1,2,4-triazole-1-ethanol, PESTANAL®, analytical standard