- Enteric Glia Regulate Gastrointestinal Motility but Are Not Required for Maintenance of the Epithelium in Mice.
Enteric Glia Regulate Gastrointestinal Motility but Are Not Required for Maintenance of the Epithelium in Mice.
When the glial fibrillary acidic protein (GFAP) promoter is used to express cellular toxins that eliminate glia in mice, intestinal epithelial permeability and proliferation increase; this led to the concept that glia are required for maintenance of the gastrointestinal epithelium. Many enteric glia, however, particularly in the mucosa, do not express GFAP. In contrast, virtually all enteric glia express proteolipid protein 1 (PLP1). We investigated whether elimination of PLP1-expressing cells compromises epithelial maintenance or gastrointestinal motility. We generated mice that express tamoxifen-inducible Cre recombinase under control of the Plp1 promoter and carry the diptheria toxin subunit A (DTA) transgene in the Rosa26 locus (Plp1 Expression of DTA in PLP1-expressing cells selectively eliminated enteric glia from the small and large intestines, but caused no defects in epithelial proliferation, barrier integrity, or ultrastructure. In contrast, administration of ganciclovir to Gfap Using the Plp1 promoter to selectively eliminate glia in mice, we found that enteric glia are not required for maintenance of the intestinal epithelium, but are required for regulation of intestinal motility in females. Previous observations supporting the concept that maintenance of the intestinal epithelium requires enteric glia can be attributed to non-glial toxicity in Gfap