MilliporeSigma
miR-181a Inhibits Cervical Cancer Development via Downregulating GRP78.
Oncology research (2017-03-01)
Chengyan Luo, Jiangnan Qiu
ABSTRACT
Cervical cancer is among the most common cancers inflicting women worldwide. Understanding the pathological mechanisms of cervical cancer development is critical for identifying novel targets for cervical cancer treatment. MicroRNAs (miRs) have various roles in regulating cancer development. In this study, we investigated the potential role of miR-181a and its target in regulating cervical cancer development and chemotherapy resistance. The expression of miR-181a was evaluated and modulated in several human cervical cancer cell lines. The role of miR-181a in regulating cervical cancer growth and chemotherapy sensitivity was investigated in cell culture models and mouse tumor xenograft models. The target of miR-181a and its function were identified in cervical cancer models. We found a distinct expression profile for miR-181a in cervical cancer cell lines. Low expression of miR-181a was closely related to cervical cancer growth and oxaliplatin resistance. HSPA5/GRP78 was identified as a target of miR-181a in cervical cancer cells. Upregulation of GRP78 led to a high cell proliferation rate and oxaliplatin resistance in cervical cancer models. In a retrospective cervical cancer cohort, high GRP78 expression was correlated with poor survival. miR-181a suppressed cervical cancer development via downregulating GRP78. High expression of GRP78 is a tumor-promoting factor in cervical cancer and is thus a potential target for novel treatment.
Sigma-Aldrich
MISSION® esiRNA, targeting human HSPA5