MilliporeSigma
  • Homochiral metal-organic frameworks based on amino acid ligands for HPLC separation of enantiomers.
Homochiral metal-organic frameworks based on amino acid ligands for HPLC separation of enantiomers.
Electrophoresis (2017-07-06)
Jun-Hui Zhang, Rui-Yu Nong, Sheng-Ming Xie, Bang-Jin Wang, Ping Ai, Li-Ming Yuan
ABSTRACT
Natural amino acids are well known to form coordination polymers with transition metal ions. In this study, six homochiral metal-organic frameworks constructed from Zn
Sigma-Aldrich
1,2-Di(4-pyridyl)ethylene, 97%