MilliporeSigma
  • Nutrients and natural toxic substances in commonly consumed Jerusalem artichoke (Helianthus tuberosus L.) tuber.
Nutrients and natural toxic substances in commonly consumed Jerusalem artichoke (Helianthus tuberosus L.) tuber.
Food chemistry (2017-09-05)
Kunchit Judprasong, Nidthida Archeepsudcharit, Kedsiri Chantapiriyapoon, Pharrunrat Tanaviyutpakdee, Piya Temviriyanukul
ABSTRACT
This study determined nutrients, chemical contaminants, (insecticide residues and heavy metals), and natural toxic substances (nitrate, nitrite, cyanide, oxalate, phytate, and trypsin inhibitor) in tubers of Jerusalem artichokes-Kaentawan in the Thai language-grown in four major provinces in Thailand. They were purchased, prepared, homogenized, and freeze-dried for further analysis using standard methods. All Kaentawan samples contained considerable amounts of fructans and dietary fiber (15.4±0.2gand3.2±0.8g/100gfresh weight [FW], respectively), as well as potassium and iron (339±61and0.32±0.05mg/100gFW, respectively). All samples had very low amounts of insecticide residues (37 compounds), cyanide, and trypsin inhibitor, as well as Pb, Cd, nitrate, and nitrite (0.82±0.09, 0.10±0.02, 1.9-17.5, and 0.01-0.24mgkg
Sigma-Aldrich
2,2-Dimethylhexane, 98%
Sigma-Aldrich
2-Methylheptane, 98%