MilliporeSigma
  • Preparation, characterization, nanostructures and bio functional analysis of sonicated protein co-precipitates from brewers' spent grain and soybean flour.
Preparation, characterization, nanostructures and bio functional analysis of sonicated protein co-precipitates from brewers' spent grain and soybean flour.
Food chemistry (2017-09-28)
Muhammad H Alu'datt, Sana Gammoh, Taha Rababah, Mohammed Almomani, Mohammad N Alhamad, Khalil Ereifej, Ali Almajwal, Asma Tahat, Neveen M Hussein, Sura Abou Nasser
ABSTRACT
This investigation was performed to assess the effects of sonication on the structure of protein, extractability of phenolics, and biological properties of isolated proteins and protein co-precipitates prepared from brewers' spent grain and soybean flour. Scanning electron micrographs revealed that the sonicated protein isolates and co-precipitates had different microstructures with fewer aggregates and smaller particles down to the nanometer scale compared to non-sonicated samples. However, the levels of free and bound phenolics extracted from non-sonicated protein isolates and protein co-precipitates increased compared to sonicated samples. The bound phenolics extracted after acid hydrolysis of sonicated protein co-precipitates showed improved ACE inhibitory activity and diminished antioxidant potency compared to non-sonicated samples. However, the free phenolics extracted from sonicated protein co-precipitates showed decreased ACE inhibitory activity and increased antioxidant activities compared to non-sonicated samples. The free and bound phenolics extracted from sonicated protein co-precipitates showed increased alpha-amylase inhibitory activity compared to non-sonicated samples.
Sigma-Aldrich
α-Amylase from hog pancreas, powder, ~50 U/mg