MilliporeSigma
  • Brief Novel Visual Experience Fundamentally Changes Synaptic Plasticity in the Mouse Visual Cortex.
Brief Novel Visual Experience Fundamentally Changes Synaptic Plasticity in the Mouse Visual Cortex.
The Journal of neuroscience : the official journal of the Society for Neuroscience (2017-08-20)
Shuo Li, Laijian Wang, Xiaoxiu Tie, Kazuhiro Sohya, Xian Lin, Alfredo Kirkwood, Bin Jiang
ABSTRACT
LTP has been known to be a mechanism by which experience modifies synaptic responses in the neocortex. Visual deprivation in the form of dark exposure or dark rearing from birth enhances NMDAR-dependent LTP in layer 2/3 of visual cortex, a process often termed metaplasticity, which may involve changes in NMDAR subunit composition and function. However, the effects of reexposure to light after dark rearing from birth on LTP induction have not been explored. Here, we showed that the light exposure after dark rearing revealed a novel NMDAR independent form of LTP in the layer 2/3 pyramidal cells in visual cortex of mice of both sexes, which is dependent on mGluR5 activation and is associated with intracellular Ca
Sigma-Aldrich
Bisindolylmaleimide II, ≥97% (Mixture of 2 isomers)
Sigma-Aldrich
6-Methyl-2-(phenylethynyl)pyridine hydrochloride, ≥98% (HPLC)