MilliporeSigma
  • Role of the Transforming Growth Factor-β in regulating hepatocellular carcinoma oxidative metabolism.
Role of the Transforming Growth Factor-β in regulating hepatocellular carcinoma oxidative metabolism.
Scientific reports (2017-10-04)
Jitka Soukupova, Andrea Malfettone, Petra Hyroššová, María-Isabel Hernández-Alvarez, Irene Peñuelas-Haro, Esther Bertran, Alexandra Junza, Jordi Capellades, Gianluigi Giannelli, Oscar Yanes, Antonio Zorzano, José Carlos Perales, Isabel Fabregat
ABSTRACT
Transforming Growth Factor beta (TGF-β) induces tumor cell migration and invasion. However, its role in inducing metabolic reprogramming is poorly understood. Here we analyzed the metabolic profile of hepatocellular carcinoma (HCC) cells that show differences in TGF-β expression. Oxygen consumption rate (OCR), extracellular acidification rate (ECAR), metabolomics and transcriptomics were performed. Results indicated that the switch from an epithelial to a mesenchymal/migratory phenotype in HCC cells is characterized by reduced mitochondrial respiration, without significant differences in glycolytic activity. Concomitantly, enhanced glutamine anaplerosis and biosynthetic use of TCA metabolites were proved through analysis of metabolite levels, as well as metabolic fluxes from U-13C6-Glucose and U-13C5-Glutamine. This correlated with increase in glutaminase 1 (GLS1) expression, whose inhibition reduced cell migration. Experiments where TGF-β function was activated with extracellular TGF-β1 or inhibited through TGF-β receptor I silencing showed that TGF-β induces a switch from oxidative metabolism, coincident with a decrease in OCR and the upregulation of glutamine transporter Solute Carrier Family 7 Member 5 (SLC7A5) and GLS1. TGF-β also regulated the expression of key genes involved in the flux of glycolytic intermediates and fatty acid metabolism. Together, these results indicate that autocrine activation of the TGF-β pathway regulates oxidative metabolism in HCC cells.
Sigma-Aldrich
BPTES, ≥95% (HPLC)