MilliporeSigma
  • Androgen regulation of signaling pathways in late fetal mouse lung development.
Androgen regulation of signaling pathways in late fetal mouse lung development.
Endocrinology (2000-08-05)
C E Dammann, S M Ramadurai, D D McCants, L D Pham, H C Nielsen
ABSTRACT
During lung development there is tension between positive and negative regulators of fibroblast-epithelial communication controlling type II cell differentiation. A clinical consequence of imbalance of this tension is the increased risk for respiratory distress syndrome in male infants. We hypothesized that chronic intrauterine androgen exposure alters fetal lung fibroblast maturation by down-regulating epidermal growth factor receptor (EGF-R) activity and by up-regulating transforming growth factor-beta receptor (TGFbeta-R) activity, leading to an inhibition of surfactant protein B (SP-B) and -C (SP-C) gene expression in type II cells. We treated pregnant mice with dihydrotestosterone (DHT; 2 mg/day) or vehicle for 7 days, starting on gestational day 11. On day 18, EGF binding, EGF-R phosphorylation, TGFbeta-R binding, and TGFbeta1-induced cell proliferation were studied in sex-specific fibroblast cultures. SP-B and -C messenger RNA levels were measured in whole lungs. Chronic DHT treatment reduced both EGF binding (females to 78+/-8% and males to 65+/-9% of controls) and EGF-induced EGF-R phosphorylation. TGFbeta-R binding was increased (females to 173+/-39% and males to 280+/-64% of controls), and TGFbeta-induced cell proliferation was increased in female cells (231+/-57% of controls). SP-B and -C messenger RNA expression was reduced to 55+/-10% and 75+/-4%, respectively. We conclude that chronic DHT exposure beginning early in lung development alters the balance of growth factor signaling that regulates lung maturation.