MilliporeSigma
  • Frequency of endometrial cancer and atypical hyperplasia in infertile women undergoing hysteroscopic polypectomy.
Frequency of endometrial cancer and atypical hyperplasia in infertile women undergoing hysteroscopic polypectomy.
The journal of obstetrics and gynaecology research (2017-07-15)
Yasushi Kuribayashi, Koji Nakagawa, Rie Sugiyama, Hiroshi Motoyama, Rikikazu Sugiyama
ABSTRACT
We aimed to determine the frequency of endometrial cancer in infertile women undergoing hysteroscopic endometrial polypectomy for endometrial polyps. A total of 1035 infertile patients who underwent office-based hysteroscopic polypectomy at Sugiyama Clinic Marunouchi between July 2011 and October 2015 were eligible for this retrospective study. All patients had been diagnosed with endometrial polyps via hysterofiberscopy prior to operation, and they underwent hysteroscopic endometrial polypectomy using a resectoscope with monopolar resection. Surgical specimens were examined histopathologically. Characteristics of patients diagnosed with endometrial cancer on histopathological examination were evaluated retrospectively. The median age of patients was 32 years (range, 19-44 years). On histopathological examination, endometrial cancer was found in 10 patients (0.97%). Each histological type of endometrial cancer was represented as follows: three cases of endometrioid adenocarcinoma G1; one of endometrioid adenocarcinoma G2; two of endometrioid adenocarcinoma G3; and four of atypical endometrial hyperplasia. The median age of endometrial cancer patients was 34 years (range, 28-41 years), and the median body mass index was 21.2 kg/m Hysteroscopic polypectomy should be performed when endometrial polyps are detected on investigational screening, and surgical specimens should be checked for the presence of malignancy.
Sigma-Aldrich
Loxoprofen, solid