MilliporeSigma
  • microRNA-219-5p inhibits epithelial-mesenchymal transition and metastasis of colorectal cancer by targeting lymphoid enhancer-binding factor 1.
microRNA-219-5p inhibits epithelial-mesenchymal transition and metastasis of colorectal cancer by targeting lymphoid enhancer-binding factor 1.
Cancer science (2017-08-05)
Lan-Xuan Huang, Chun-Yan Hu, Li Jing, Min-Cong Wang, Meng Xu, Jing Wang, Yu Wang, Ke-Jun Nan, Shu-Hong Wang
ABSTRACT
Aberrant expression of microRNAs (miRs) has been shown to play a critical role in the pathogenesis and progression of tumors. microRNA-219-5p (miR-219-5p) has been reported to be abnormally expressed in some types of human tumors. However, the mechanism between miR-219-5p and colorectal cancer (CRC) metastasis remains unclear. In the present study, miR-219-5p was found to be downregulated in CRC tissue compared with matched normal tissue. Through luciferase reporter assay, we demonstrated lymphoid enhancer-binding factor 1 (LEF1) as a direct target of miR-219-5p. Overexpression of miR-219-5p could inhibit motility, migration and invasion of CRC cells, and inhibit epithelial-mesenchymal transition (EMT). Furthermore, silencing LEF1 phenocopied this metastasis-suppressive function. The recovery experiment showed that re-expression of LEF1 rescued this suppressive effect on tumor metastasis and reversed the expression of EMT markers caused by miR-219-5p. Additionally, we demonstrated that miR-219-5p exerted this tumor-suppressive function by blocking activation of the AKT and ERK pathways. Finally, a nude mice experiment showed that miR-219-5p reduced the lung metastasis ability of CRC cells. Taken together, our findings indicate that miR-219-5p inhibits metastasis and EMT of CRC by targeting LEF1 and suppressing the AKT and ERK pathways, which may provide a new antitumor strategy to delay CRC metastasis.
Sigma-Aldrich
MISSION® esiRNA, targeting human LEF1