MilliporeSigma
  • The Nrf2/GCH1/BH4 Axis Ameliorates Radiation-Induced Skin Injury by Modulating the ROS Cascade.
The Nrf2/GCH1/BH4 Axis Ameliorates Radiation-Induced Skin Injury by Modulating the ROS Cascade.
The Journal of investigative dermatology (2017-06-10)
Jiao Xue, Chenxiao Yu, Wenjiong Sheng, Wei Zhu, Judong Luo, Qi Zhang, Hongying Yang, Han Cao, Wenjie Wang, Jundong Zhou, Jinchang Wu, Peng Cao, Ming Chen, Wei-Qun Ding, Jianping Cao, Shuyu Zhang
ABSTRACT
Radiation-induced skin injury is a common side effect of radiotherapy and can limit the duration and dose of radiotherapy. Most early work focused on elimination of reactive oxygen species (ROS) after radiation; however, less is known about the mechanisms underlying amplification of ROS and consequent skin injury by radiation. 5,6,7,8-Tetrahydrobiopterin (BH4) is an essential cofactor for all nitric oxide synthases. Inadequate availability of BH4 leads to uncoupling of nitric oxide synthases and production of highly oxidative radicals. In this study, we demonstrated that radiation disrupted BH4, which resulted in nitric oxide synthases uncoupling and augmented radiation-induced ROS. Overexpression of GTP cyclohydrolase I (GCH1), the rate-limiting enzyme for BH4 synthesis, restored cellular BH4 levels and nitric oxide production and decreased radiation-induced ROS. GCH1 also protected skin cells and rat skins against radiation-induced damage. We found that GCH1 was regulated by NF-E2-related factor 2, a key mediator of the cellular antioxidant response. Importantly, we identified GCH1 as a key effector for NF-E2-related factor 2-mediated protection against radiation-induced skin injury by inhibiting ROS production. Taken together, the findings of this study illustrate the key role of the NF-E2-related factor 2/GCH1/BH4 axis during radiation-induced skin damage.
Sigma-Aldrich
MISSION® esiRNA, targeting human GCH1
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder