MilliporeSigma
  • EZH2 and BCL6 Cooperate to Assemble CBX8-BCOR Complex to Repress Bivalent Promoters, Mediate Germinal Center Formation and Lymphomagenesis.
EZH2 and BCL6 Cooperate to Assemble CBX8-BCOR Complex to Repress Bivalent Promoters, Mediate Germinal Center Formation and Lymphomagenesis.
Cancer cell (2016-08-10)
Wendy Béguelin, Matt Teater, Micah D Gearhart, María Teresa Calvo Fernández, Rebecca L Goldstein, Mariano G Cárdenas, Katerina Hatzi, Monica Rosen, Hao Shen, Connie M Corcoran, Michelle Y Hamline, Randy D Gascoyne, Ross L Levine, Omar Abdel-Wahab, Jonathan D Licht, Rita Shaknovich, Olivier Elemento, Vivian J Bardwell, Ari M Melnick
ABSTRACT
The EZH2 histone methyltransferase mediates the humoral immune response and drives lymphomagenesis through formation of bivalent chromatin domains at critical germinal center (GC) B cell promoters. Herein we show that the actions of EZH2 in driving GC formation and lymphoma precursor lesions require site-specific binding by the BCL6 transcriptional repressor and the presence of a non-canonical PRC1-BCOR-CBX8 complex. The chromodomain protein CBX8 is induced in GC B cells, binds to H3K27me3 at bivalent promoters, and is required for stable association of the complex and the resulting histone modifications. Moreover, oncogenic BCL6 and EZH2 cooperate to accelerate diffuse large B cell lymphoma (DLBCL) development and combinatorial targeting of these repressors results in enhanced anti-lymphoma activity in DLBCLs.
Sigma-Aldrich
FX1, ≥98% (HPLC)