MilliporeSigma
  • Structural Determinants for the Selectivity of the Positive KCa3.1 Gating Modulator 5-Methylnaphtho[2,1-
Structural Determinants for the Selectivity of the Positive KCa3.1 Gating Modulator 5-Methylnaphtho[2,1-
Molecular pharmacology (2017-08-02)
Brandon M Brown, Heesung Shim, Miao Zhang, Vladimir Yarov-Yarovoy, Heike Wulff
ABSTRACT
Intermediate-conductance (K
Sigma-Aldrich
NS309, ≥98% (HPLC), solid
Sigma-Aldrich
SKA-121, ≥98% (HPLC)