MilliporeSigma
Regulation of hypoxia-induced autophagy in glioblastoma involves ATG9A.
British journal of cancer (2017-08-11)
Siti Aminah Abdul Rahim, Anne Dirkse, Anais Oudin, Anne Schuster, Jill Bohler, Vanessa Barthelemy, Arnaud Muller, Laurent Vallar, Bassam Janji, Anna Golebiewska, Simone P Niclou
ABSTRACT
Hypoxia is negatively associated with glioblastoma (GBM) patient survival and contributes to tumour resistance. Anti-angiogenic therapy in GBM further increases hypoxia and activates survival pathways. The aim of this study was to determine the role of hypoxia-induced autophagy in GBM. Pharmacological inhibition of autophagy was applied in combination with bevacizumab in GBM patient-derived xenografts (PDXs). Sensitivity towards inhibitors was further tested in vitro under normoxia and hypoxia, followed by transcriptomic analysis. Genetic interference was done using ATG9A-depleted cells. We find that GBM cells activate autophagy as a survival mechanism to hypoxia, although basic autophagy appears active under normoxic conditions. Although single agent chloroquine treatment in vivo significantly increased survival of PDXs, the combination with bevacizumab resulted in a synergistic effect at low non-effective chloroquine dose. ATG9A was consistently induced by hypoxia, and silencing of ATG9A led to decreased proliferation in vitro and delayed tumour growth in vivo. Hypoxia-induced activation of autophagy was compromised upon ATG9A depletion. This work shows that inhibition of autophagy is a promising strategy against GBM and identifies ATG9 as a novel target in hypoxia-induced autophagy. Combination with hypoxia-inducing agents may provide benefit by allowing to decrease the effective dose of autophagy inhibitors.
Sigma-Aldrich
MISSION® esiRNA, targeting human ATG9A
Pricing and availability is not currently available.