Skip to Content
CA
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
mRNA Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Home
Search Results
m
m
Nature (2017-09-05)
Chunxia Zhang, Yusheng Chen, Baofa Sun, Lu Wang, Ying Yang, Dongyuan Ma, Junhua Lv, Jian Heng, Yanyan Ding, Yuanyuan Xue, Xinyan Lu, Wen Xiao, Yun-Gui Yang, Feng Liu
PMID
28869969
ABSTRACT
N
Product Description
P9424
Millipore
ProteinA-Sepharose
™
4, Fast Flow from
Staphylococcus aureus
, aqueous ethanol suspension
Pricing
EHU063661
Sigma-Aldrich
MISSION
®
esiRNA, targeting human METTL3
Pricing
SML0649
Sigma-Aldrich
Deshydroxy LY-411575, ≥98% (HPLC)
Pricing