MilliporeSigma
  • Increased expression of latent TGF-β-binding protein 4 affects the fibrotic process in scleroderma by TGF-β/SMAD signaling.
Increased expression of latent TGF-β-binding protein 4 affects the fibrotic process in scleroderma by TGF-β/SMAD signaling.
Laboratory investigation; a journal of technical methods and pathology (2017-03-07)
Jiaying Lu, Qingmei Liu, Lei Wang, Wenzhen Tu, Haiyan Chu, Weifeng Ding, Shuai Jiang, Yanyun Ma, Xiangguang Shi, Weilin Pu, Xiaodong Zhou, Li Jin, Jiucun Wang, Wenyu Wu
ABSTRACT
Scleroderma is a fibrosis-related disorder characterized by cutaneous and internal organ fibrosis, and excessive collagen deposition in extracellular matrix (ECM) is a major cause of fibrosis. Transforming growth factor-β (TGF-β)/SMAD signaling has a central role in the pathogenesis of fibrosis by inducing abnormal collagen accumulation in ECM, and latent TGF-β-binding protein 4 (LTBP-4) affects the secretion of latent TGF-β to ECM. A previous study indicated that bleomycin (BLM) treatment increased LTBP-4 expression in lung fibroblasts of Thy-1 knockout mice with lung fibrosis, and LTBP-4 further promoted TGF-β bioavailability as well as SMAD3 phosphorylation. However, the expression and function of LTBP-4 in human scleroderma remain unclear. We aimed to investigate the potential role of LTBP-4 in scleroderma through clinical, in vivo and in vitro studies. LTBP-4 and TGF-β expressions were significantly upregulated in systemic scleroderma (SSc) patients' plasma compared with normal controls (LTBP-4, 1,215±100.2 vs 542.8±41.7 ng/ml, P<0.0001; TGF-β, 1.5±0.2 vs 0.7±0.1 ng/ml, P=0.0031), while no significant difference was found between localized scleroderma (LSc) and normal controls. The plasma concentrations of LTBP-4 and TGF-β were even higher in SSc patients with lung fibrosis (LTBP-4, 1462± 137.3 vs 892.8±113.4 ng/ml, P=0.0037; TGF-β, 2.0±0.4 vs 0.9±0.2 ng/ml, P=0.0212) and esophagus involvement (1390±134.4 vs 940.7±127.0 ng/ml, P=0.0269; TGF-β, 1.9±0.3 vs 0.9±0.2 ng/ml, P=0.0426). The area under receiver operating characteristics (ROC) curve of LTBP-4 was 0.86. Immunohistochemistry measurement also demonstrated a higher LTBP-4 expression in sclerotic skin tissue of LSc and SSc compared with normal controls. More positive fibroblasts were also found in BLM-induced scleroderma mouse model than the saline-treated group. In in vitro studies, knockdown of LTBP-4 in SSc skin fibroblasts prominently reduced downstream COL1A1, COL1A2, and COL3A1 mRNA level by 84%, 82%, and 43%, respectively, and other fibrosis-related genes' expression were also decreased. Furthermore, extracellular TGF-β level and the SMAD2/3 phosphorylation were inhibited through LTBP-4 knockdown treatment, suggesting that the knockdown of LTBP-4 reduced the collagen expression through TGF-β/SMAD signaling pathway. Taken together, these data suggest that LTBP-4 affects fibrotic process in scleroderma, and the high expression of LTBP-4 in SSc plasma may serve as a clinical biomarker in diagnosing this disease. In addition, this study also lays the theoretical foundation for targeting LTBP-4 as treatment of scleroderma.
Sigma-Aldrich
MISSION® esiRNA, targeting human LTBP4