MilliporeSigma
Self-assembly of zinc hydroxystannate on amorphous hydrous TiO
Journal of hazardous materials (2017-07-18)
Wei Wang, Yongchun Kan, Jiajia Liu, Kim Meow Liew, Lei Liu, Yuan Hu
ABSTRACT
Zinc hydroxystannate (ZHS) was fabricated on the surface of amorphous hydrous TiO