MilliporeSigma
  • Hydrophilic interaction chromatography with a focus on the drug-phosphate interaction in drug screening to determine the phospholipidosis induction risk.
Hydrophilic interaction chromatography with a focus on the drug-phosphate interaction in drug screening to determine the phospholipidosis induction risk.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2017-03-13)
Haruka Okamoto, Ryohei Hamaguchi, Yukihiro Kuroda
ABSTRACT
Cationic amphiphilic drugs (CADs) can induce the hyperaccumulation of phospholipids in cells and tissues. This side effect, which is known as drug-induced phospholipidosis, is sometimes problematic in the development and clinical use of CADs. It is known that CADs generally interact with phospholipids via both hydrophobic and acid-base interactions, and CADs with the larger affinity to phospholipid exhibit the larger induction risk. To develop a chromatographic assay system to predict the phospholipidosis-inducing potential with considering the acid-base interaction between CAD and phosphate group of phospholipid, hydrophilic interaction chromatographic (HILIC) methods were tested in this study. First, a PC HILIC column with phosphocholine groups on a packed material was used. The acid-base or other hydrophilic interactions to the stationary phase differed among basic drugs, and retention to the PC HILIC column did not accurately reflect the induction potential of phospholipidosis. As an alternative HILIC approach, the elution of CADs with the phosphate buffer from an amide column was tested. The elution effect, which is expressed as ratio of retention factors between different phosphate content in the mobile phase, closely correlated with the induction potential. Using the elution effect and retention factor to a reversed-phase HPLC column, the phospholipidosis-inducing drugs were clearly discriminated from the non-inducers. These results suggest that the proposed chromatographic approach can screen phospholipidosis-inducing drugs.
Sigma-Aldrich
Pindolol, ≥98% (TLC), powder
Sigma-Aldrich
Amiodarone hydrochloride, ≥98%