MilliporeSigma
  • Deletion of 12/15-lipoxygenase accelerates the development of aging-associated and instability-induced osteoarthritis.
Deletion of 12/15-lipoxygenase accelerates the development of aging-associated and instability-induced osteoarthritis.
Osteoarthritis and cartilage (2017-07-12)
L Habouri, F E El Mansouri, Y Ouhaddi, B Lussier, J-P Pelletier, J Martel-Pelletier, M Benderdour, H Fahmi
ABSTRACT
12/15-Lipoxygenase (12/15-LOX) catalyzes the generation of various anti-inflammatory lipid mediators, and has been implicated in several inflammatory and degenerative diseases. However, there is currently no evidence that 12/15-LOX has a role in osteoarthritis (OA). The aim of this study was to investigate the role of 12/15-LOX in the pathogenesis of OA. The development of aging-associated and destabilization of the medial meniscus (DMM)-induced OA were compared in 12/15-LOX-deficient (12/15-LOX-/-) and wild-type (WT) mice. The extent of cartilage damage was evaluated by histology. The expression of OA markers was evaluated by immunohistochemistry and RT-PCR. Cartilage explants were stimulated with IL-1α in the absence or presence of the 12/15-LOX metabolites, 15-hydroxyeicosatetraenoic acids (15-HETE), 13-hydroxyoctadecadienoic acid (13-HODE) or lipoxin A4 (LXA4), and the levels of matrix metalloproteinases-13 (MMP-13), Nitric oxide (NO) and prostaglandin E The expression of 12/15-LOX in cartilage increased during the progression of DMM-induced OA and with aging in WT mice. Cartilage degeneration was more severe in 12/15-LOX-/- mice compared to WT mice in both models of OA, and this was associated with increased expression of MMP-13, a disintegrin and metalloproteinase with thrombospondin motifs, aggrecanases (ADAMTS5), inducible NO synthases (iNOS), and mPGES-1. Treatment of cartilage explants with 12/15-LOX metabolites, suppressed IL-1α-induced production of MMP-13, NO and PGE These data suggest an important role of 12/15-LOX in the pathogenesis of OA. They also suggest that activation of this pathway may provide a novel strategy for prevention and treatment of OA.
Sigma-Aldrich
Lipoxin A4, ethanol solution