MilliporeSigma
  • Scaffold of Selenium Nanovectors and Honey Phytochemicals for Inhibition of
Scaffold of Selenium Nanovectors and Honey Phytochemicals for Inhibition of
Frontiers in cellular and infection microbiology (2017-04-08)
Prateeksha, Braj R Singh, M Shoeb, S Sharma, A H Naqvi, Vijai K Gupta, Brahma N Singh
ABSTRACT
Honey is an excellent source of polyphenolic compounds that are effective in attenuating quorum sensing (QS), a chemical process of cell-to-cell communication system used by the opportunistic pathogen
Sigma-Aldrich
(Z-)-4-Bromo-5-(bromomethylene)-2(5H)-furanone, ≥97.0% (GC)
Sigma-Aldrich
Pyocyanin, from Pseudomonas aeruginosa, ≥98% (HPLC)