MilliporeSigma
  • The L-type Voltage-Gated Calcium Channel co-localizes with Syntaxin 1A in nano-clusters at the plasma membrane.
The L-type Voltage-Gated Calcium Channel co-localizes with Syntaxin 1A in nano-clusters at the plasma membrane.
Scientific reports (2017-09-14)
Julia Sajman, Michael Trus, Daphne Atlas, Eilon Sherman
ABSTRACT
The secretory signal elicited by membrane depolarization traverses from the Ca
Sigma-Aldrich
Auranofin, ≥98% (HPLC)