MilliporeSigma
Molecular mechanism of mRNA repression in
The EMBO journal (2017-03-25)
Alexandre Smirnov, Chuan Wang, Lisa L Drewry, Jörg Vogel
ABSTRACT
Research into post-transcriptional control of mRNAs by small noncoding RNAs (sRNAs) in the model bacteria
Sigma-Aldrich
Anti-GroEL antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)