MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Ligand sensitized strong luminescence from Eu
Dalton transactions (Cambridge, England : 2003) (2017-07-15)
Tuhin Samanta, Sourav Kanti Jana, Athma E Praveen, Venkataramanan Mahalingam
ABSTRACT
Lanthanide (Ln)-doped nanocrystals generally display low luminescence quantum efficiency due to forbidden nature of the 4f-4f transition besides possessing low absorption cross sections (∼10 M
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
4,4,4-Trifluoro-1-phenyl-1,3-butanedione, 99%
Sigma-Aldrich
Oleic acid, technical grade, 90%