MilliporeSigma
  • Phenotypic defects in newborn Gammarus fossarum (Amphipoda) following embryonic exposure to fenoxycarb.
Phenotypic defects in newborn Gammarus fossarum (Amphipoda) following embryonic exposure to fenoxycarb.
Ecotoxicology and environmental safety (2017-06-18)
Hélène Arambourou, Arnaud Chaumot, Emmanuelle Vulliet, Gaëlle Daniele, Nicolas Delorme, Khédidja Abbaci, Vincent Debat
ABSTRACT
During morphogenesis numerous morphogenetic factors ensure the production of a target phenotype. By disrupting these processes, a toxic exposure during this period could cause an increase of phenotypic defects. In the present study, embryos of the freshwater amphipod Gammarus fossarum were exposed throughout the embryogenesis to increasing concentrations of fenoxycarb (0, 0.5µgL
Product Description
Supelco
Fenoxycarb, PESTANAL®, analytical standard