MilliporeSigma
  • Mathematical description of pH-stat kinetic traces measured during photochemical quinone decomposition.
Mathematical description of pH-stat kinetic traces measured during photochemical quinone decomposition.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology (2016-12-13)
Virág Kiss, Gábor Lehoczki, Katalin Ősz
ABSTRACT
Substituted 1,4-benzoquinone (QR) derivatives are photosensitive in aqueous solution and form hydroquinones (QR-H
Sigma-Aldrich
2,5-Dichloro-1,4-benzoquinone, 98%