MilliporeSigma
PD-L1 up-regulation restrains Th17 cell differentiation in
The Journal of experimental medicine (2017-07-16)
Yuan Zhang, Chi A Ma, Monica G Lawrence, Timothy J Break, Michael P O'Connell, Jonathan J Lyons, Diego B López, John S Barber, Yongge Zhao, Daniel L Barber, Alexandra F Freeman, Steven M Holland, Michail S Lionakis, Joshua D Milner
ABSTRACT
Patients with hypomorphic mutations in
Sigma-Aldrich
MISSION® esiRNA, targeting human SOCS3