MilliporeSigma
Identifying the charge generation dynamics in Cs
Physical chemistry chemical physics : PCCP (2017-08-16)
Manuel Salado, Ramesh K Kokal, Laura Calio, Samrana Kazim, Melepurath Deepa, Shahzada Ahmad
ABSTRACT
Triple cation based perovskite solar cells offer enhanced moisture tolerance and stability compared to mixed perovskites. Slight substitution of methyl ammonium or formamidinium cation by cesium (Cs
Sigma-Aldrich
Formamidinium iodide
Sigma-Aldrich
Methylammonium bromide