MilliporeSigma
  • Experimental Studies on the Hydrotreatment of Kraft Lignin to Aromatics and Alkylphenolics Using Economically Viable Fe-Based Catalysts.
Experimental Studies on the Hydrotreatment of Kraft Lignin to Aromatics and Alkylphenolics Using Economically Viable Fe-Based Catalysts.
ACS sustainable chemistry & engineering (2017-04-18)
Shilpa Agarwal, Ramesh Kumar Chowdari, Idoia Hita, Hero Jan Heeres
ABSTRACT
Limonite, a low-cost iron ore, was investigated as a potential hydrotreatment catalyst for kraft lignin without the use of an external solvent (batch reactor, initial H
Sigma-Aldrich
Goethite, 30-63% Fe