MilliporeSigma
  • Analytical determination of bisphenol A (BPA) and bisphenol analogues in paper products by GC-MS/MS.
Analytical determination of bisphenol A (BPA) and bisphenol analogues in paper products by GC-MS/MS.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2017-04-13)
A Jurek, E Leitner
ABSTRACT
Bisphenol A (BPA; 4-[2-(4-hydroxyphenyl)propan-2-yl]phenol), a suspected endocrine disruptor with a weak estrogenic activity, is used in a variety of consumer products, including food-contact materials made of paper and cardboard products. Due to restrictions on the use of BPA because of its potential health risks, BPA is gradually being replaced by other bisphenols because no limitations exist for these substances. This study presents a method for the simultaneous analysis of BPA, bisphenol AF (BPAF), bisphenol B (BPB), bisphenol E (BPE), bisphenol F (BPF) and bisphenol S (BPS) in paper and board products using gas chromatography-tandem mass spectrometry (GC-MS/MS). Paper samples were extracted by liquid extraction, as well as by Folch extraction, derivatised with N,O-bis(trimethylsilyl)trifluoroacetamide (BSTFA) and the results compared. The developed method showed good linearity (R
Sigma-Aldrich
4,4′-(Hexafluoroisopropylidene)diphenol, 97%
Sigma-Aldrich
4,4′-Sulfonyldiphenol, 98%
Sigma-Aldrich
Bis(4-hydroxyphenyl)methane, 98%