MilliporeSigma
  • Nitric oxide is involved in the oxytetracycline-induced suppression of root growth through inhibiting hydrogen peroxide accumulation in the root meristem.
Nitric oxide is involved in the oxytetracycline-induced suppression of root growth through inhibiting hydrogen peroxide accumulation in the root meristem.
Scientific reports (2017-02-22)
Qing-Xiang Yu, Golam Jalal Ahammed, Yan-Hong Zhou, Kai Shi, Jie Zhou, Yunlong Yu, Jing-Quan Yu, Xiao-Jian Xia
ABSTRACT
Use of antibiotic-contaminated manure in crop production poses a severe threat to soil and plant health. However, few studies have studied the mechanism by which plant development is affected by antibiotics. Here, we used microscopy, flow cytometry, gene expression analysis and fluorescent dyes to study the effects of oxytetracycline (OTC), a widely used antibiotic in agriculture, on root meristem activity and the accumulation of hydrogen peroxide (H
Sigma-Aldrich
2′,7′-Dichlorodihydrofluorescein diacetate, ≥97%
Sigma-Aldrich
Diphenyleneiodonium chloride, ≥98%