MilliporeSigma
  • Interactions of Hg(ii) with oligonucleotides having thymine-thymine mispairs. Optimization of an impedimetric Hg(ii) sensor.
Interactions of Hg(ii) with oligonucleotides having thymine-thymine mispairs. Optimization of an impedimetric Hg(ii) sensor.
The Analyst (2017-05-06)
Ajar Kamal, Zhe She, Renu Sharma, Heinz-Bernhard Kraatz
ABSTRACT
The present work describes the effect of the number of thymine-thymine mispairs in single strand DNA probes on Hg(ii) interactions and further to develop a highly sensitive DNA based impedimetric sensor for Hg(ii) detection. To achieve this goal, the influence of the number of T-T mispairs on the signal response prompted by DNA-Hg(ii) binding interactions was examined on three designed DNA probes: 5'-OH-(CH
Sigma-Aldrich
Sodium perchlorate, ACS reagent, ≥98.0%