MilliporeSigma
  • Vitamin D supplementation inhibits oxidative stress and upregulate SIRT1/AMPK/GLUT4 cascade in high glucose-treated 3T3L1 adipocytes and in adipose tissue of high fat diet-fed diabetic mice.
Vitamin D supplementation inhibits oxidative stress and upregulate SIRT1/AMPK/GLUT4 cascade in high glucose-treated 3T3L1 adipocytes and in adipose tissue of high fat diet-fed diabetic mice.
Archives of biochemistry and biophysics (2017-01-09)
Prasenjit Manna, Arunkumar E Achari, Sushil K Jain
ABSTRACT
This study examined the hypothesis that vitamin-D prevents oxidative stress and upregulates glucose metabolism via activating insulin-independent signaling molecules in 3T3-L1 adipocytes and in high fat diet (HFD)-fed mice. To investigate the mechanism 3T3L1 adipocytes were treated with high glucose (HG, 25 mM) and 1,25(OH)
Sigma-Aldrich
MISSION® esiRNA, targeting human INSR