MilliporeSigma
  • Heterogenic response of prokaryotes toward silver nanoparticles and ions is facilitated by phenotypes and attachment of silver aggregates to cell surfaces.
Heterogenic response of prokaryotes toward silver nanoparticles and ions is facilitated by phenotypes and attachment of silver aggregates to cell surfaces.
Cytometry. Part A : the journal of the International Society for Analytical Cytology (2017-01-23)
Yuting Guo, Hans J Stärk, Gerd Hause, Matthias Schmidt, Hauke Harms, Lukas Y Wick, Susann Müller
ABSTRACT
Tons of anthropogenic silver nanoparticles (AgNPs) are assumed to be released into the environment due to their use in many consumer products. AgNPs are known to be toxic toward microorganisms and thus may harm their specific functions in ecosystems. Here we explore the impact of AgNPs on functioning of single cells in microbial populations at doses typically found in anthropogenic environments. The response of single cells to AgNPs was analyzed by flow cytometry and using the fluorescent dyes propidium iodide and DiBAC
Sigma-Aldrich
TERGITOL, Type NP-10
Sigma-Aldrich
Bis(1,3-dibutylbarbituric acid) trimethine oxonol, ≥95% (HPLC)