MilliporeSigma
  • Shaping nanofiltration channels in a carbonaceous membrane via controlling the pyrolysis atmosphere.
Shaping nanofiltration channels in a carbonaceous membrane via controlling the pyrolysis atmosphere.
Physical chemistry chemical physics : PCCP (2017-08-02)
Zheng Xing, Yeap Hung Ng, Siok-Wei Tay, Rachel Pek Har Oon, Liang Hong
ABSTRACT
This work investigates the effect of atmosphere on pyrolysis of a polymer matrix (precursor) for directing its transformation towards more disordered graphene species and smaller graphitic nanograins. These two structural characteristics are crucial to the generation of nano-channels (NCs) pertinent to nanofiltration (NF). Two measures are explored hereby to conduct the study: varying the pyrolysis atmosphere and implementing highly dispersed nickel atomic clusters (Ni-clusters) in the coating matrix undergoing pyrolysis. A thermally reactive polymer precursor is developed to allow the above two measures to act more effectively. The various pyrolysis atmospheres employed include inert Ar, a reducing H
Sigma-Aldrich
Lauryl methacrylate, contains 500 ppm MEHQ as inhibitor, 96%
Sigma-Aldrich
Dodecylamine, 98%