MilliporeSigma
  • Quantification of synthetic cannabinoids in herbal smoking blends using NMR.
Quantification of synthetic cannabinoids in herbal smoking blends using NMR.
Drug testing and analysis (2016-07-13)
Simon J Dunne, Jenny P Rosengren-Holmberg
ABSTRACT
Herbal smoking blends containing synthetic cannabinoids have become popular alternatives to marijuana. These products were previously sold in pre-packaged foil bags, but nowadays seizures usually contain synthetic cannabinoid powders together with unprepared plant materials. A question often raised by the Swedish police is how much smoking blend can be prepared from certain amounts of banned substance, in order to establish the severity of the crime. To address this question, information about the synthetic cannabinoid content in both the powder and the prepared herbal blends is necessary. In this work, an extraction procedure compatible with direct NMR quantification of synthetic cannabinoids in herbal smoking blends was developed. Extraction media, time and efficiency were tested for different carrier materials containing representative synthetic cannabinoids. The developed protocol utilizes a 30 min extraction step in d
Sigma-Aldrich
Tridecane, ≥99%
Sigma-Aldrich
1,3,5-Trimethoxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
L-Valine ethyl ester hydrochloride, 99%