MilliporeSigma
  • Cytohesin 1 regulates homing and engraftment of human hematopoietic stem and progenitor cells.
Cytohesin 1 regulates homing and engraftment of human hematopoietic stem and progenitor cells.
Blood (2016-12-03)
Justyna Rak, Katie Foster, Katarzyna Potrzebowska, Mehrnaz Safaee Talkhoncheh, Natsumi Miharada, Karolina Komorowska, Therese Torngren, Anders Kvist, Åke Borg, Lena Svensson, Dominique Bonnet, Jonas Larsson
ABSTRACT
Adhesion is a key component of hematopoietic stem cell regulation mediating homing and retention to the niche in the bone marrow. Here, using an RNA interference screen, we identify cytohesin 1 (CYTH1) as a critical mediator of adhesive properties in primary human cord blood-derived hematopoietic stem and progenitor cells (HSPCs). Knockdown of CYTH1 disrupted adhesion of HSPCs to primary human mesenchymal stroma cells. Attachment to fibronectin and ICAM1, 2 integrin ligands, was severely impaired, and CYTH1-deficient cells showed a reduced integrin β1 activation response, suggesting that CYTH1 mediates integrin-dependent functions. Transplantation of CYTH1-knockdown cells to immunodeficient mice resulted in significantly lower long-term engraftment levels, associated with a reduced capacity of the transplanted cells to home to the bone marrow. Intravital microscopy showed that CYTH1 deficiency profoundly affects HSPC mobility and localization within the marrow space and thereby impairs proper lodgment into the niche. Thus, CYTH1 is a novel major regulator of adhesion and engraftment in human HSPCs through mechanisms that, at least in part, involve the activation of integrins.
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder